accessibility menu, dialog, popup

UserName

CAS RN 26530-20-1

IUPAC Name: 2-octyl-2,3-dihydro-1,2-thiazol-3-one
Synonyms: octylisothiazolinone
Molecular Weight (g/mol): 213.34
Molecular Formula: C11H19NOS
InChi Key: JPMIIZHYYWMHDT-UHFFFAOYSA-N
SMILES: CCCCCCCCN1SC=CC1=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results