missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ 2-Octyl-4-isothiazolin-3-one, MilliporeSigma™ Supelco™

Catalog No. 111018773
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018773 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018773 Supplier Merck Emd Millipore Supplier No. 46078250MGR
Agriculture

Specifications

Product Line PESTANAL
CAS 26530-20-1
Physical Form Neat
Linear Formula C11H19 NOS
MDL Number MFCD00072473
Synonym 2-Octyl-3(2 H)-isothiazolone; Octhilinone
Type Analytical Standard
Quantity 250 mg
Formula Weight 213.34
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH