accessibility menu, dialog, popup

UserName

HOOBr

CAS RN 2623-87-2

IUPAC Name: 4-bromobutanoic acid
Synonyms: 4-bromobutanoic acid
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: GRHQDJDRGZFIPO-UHFFFAOYSA-N
SMILES: OC(=O)CCCBr
Boiling Point 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)