missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyric acid, 97%

Catalog No. AAA1287030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1287030 250 g
AAA1287009 10 g
AAA1287018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1287030 Supplier Thermo Scientific Chemicals Supplier No. A1287030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2623-87-2 | C4H7BrO2 | 167.00 g/mol

4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.

Solubility
Soluble in water and dilute alkali.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to light. Incompatible with strong oxidizing agents

Chemical Identifiers

CAS 2623-87-2
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00002817
InChI Key GRHQDJDRGZFIPO-UHFFFAOYSA-N
Synonym 4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754
PubChem CID 75809
IUPAC Name 4-bromobutanoic acid
SMILES OC(=O)CCCBr

Specifications

Melting Point 31°C to 33°C
Boiling Point 129°C to 131°C (11 mmHg)
Flash Point 115°C (239°F)
Odor Stench
Quantity 250 g
UN Number UN3261
Beilstein 1743056
Sensitivity Light sensitive
Solubility Information Soluble in water and dilute alkali.
Formula Weight 167.01
Percent Purity 97%
Chemical Name or Material 4-Bromobutyric acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-083-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only