missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyric Acid 95.0+%, TCI America™

Catalog No. B127925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B127925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B127925G Supplier TCI America Supplier No. B127925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2623-87-2
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00002817
InChI Key GRHQDJDRGZFIPO-UHFFFAOYSA-N
Synonym 4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754
PubChem CID 75809
IUPAC Name 4-bromobutanoic acid
SMILES OC(=O)CCCBr

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 142°C
Quantity 25 g
UN Number 3261
Formula Weight 167.00
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromobutyric Acid