accessibility menu, dialog, popup

UserName

CAS RN 2615-25-0

IUPAC Name: cyclohexane-1,4-diamine
Synonyms: 1,4-diaminocyclohexane
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: VKIRRGRTJUUZHS-UHFFFAOYSA-N
SMILES: NC1CCC(N)CC1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)