missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Diaminocyclohexane, 98+%

Catalog No. AC406361000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406361000 100 g Plastic bottle
AC406360250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406361000 Supplier Thermo Scientific Chemicals Supplier No. 406361000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2615-25-0 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2615-25-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00035862,MFCD00075174,MFCD08276323
InChI Key VKIRRGRTJUUZHS-UHFFFAOYSA-N
Synonym trans-1,4-diaminocyclohexane, trans-cyclohexane-1,4-diamine, 1,4-diaminocyclohexane, trans-1,4-cyclohexanediamine, cis-cyclohexane-1,4-diamine, 1,4-cyclohexanediamine, cis-1,4-cyclohexanediamine, 1,4-cyclohexanediamine, trans, 1r,4r-cyclohexane-1,4-diamine, 1,4-cyclohexanediamine, cis
PubChem CID 18374
IUPAC Name cyclohexane-1,4-diamine
SMILES NC1CCC(N)CC1

Specifications

Melting Point 68.0°C to 71.0°C
Color Brown to White
Boiling Point 197.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H10(NH2)2
Quantity 100 g
Formula Weight 114.19
Percent Purity 99%
Physical Form Crystals and/or Chunks
Chemical Name or Material trans-1, 4-Diaminocyclohexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only