accessibility menu, dialog, popup

UserName

CAS RN 2610-10-8

IUPAC Name: hexasodium (3E)-4-oxo-7-({[(6E)-5-oxo-7-sulfonato-6-(2-{2-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-5,6-dihydronaphthalen-2-yl]carbamoyl}amino)-3-(2-{2-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2-sulfonate
Synonyms: hexasodium (3E)-4-oxo-7-({[(6E)-5-oxo-7-sulfonato-6-(2-{2-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)naphthalen-2-yl]carbamoyl}amino)-3-(2-{2-sulfonato-4-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)naphthalene-2-sulfonate
Molecular Weight (g/mol): 1373.05
Molecular Formula: C45H26N10Na6O21S6
InChi Key: LDCKXVAPQFUIOI-ZFLDVXHKSA-H
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C3=CC=C(NC(=O)NC4=CC=C5C(=C4)C=C(\C(=N\NC4=CC=C(C=C4S([O-])(=O)=O)N=NC4=CC=C(C=C4)S([O-])(=O)=O)C5=O)S([O-])(=O)=O)C=C3C=C2S([O-])(=O)=O)C(=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results