missing translation for 'onlineSavingsMsg'
Learn More

Direct Red 80 [for Biochemical Research], TCI America™

Catalog No. D41325G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41325G Supplier TCI America Supplier No. D41325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2610-10-8
Molecular Formula C45H26N10Na6O21S6
Molecular Weight (g/mol) 1373.05
MDL Number MFCD00054389
InChI Key LDCKXVAPQFUIOI-ZFLDVXHKSA-H
Synonym hexasodium 4-hydroxy-7-5-hydroxy-7-sulfonato-6-e-2-2-sulfonato-4-e-2-4-sulfonatophenyl diazen-1-yl phenyl diazen-1-yl naphthalen-2-yl carbamoyl amino-3-e-2-2-sulfonato-4-e-2-4-sulfonatophenyl diazen-1-yl phenyl diazen-1-yl naphthalene-2-sulfonate, 2-naphthalenesulfonic acid, 7,7'-carbonyldiimino bis 4-hydroxy-3-2-2-sulfo-4-2-4-sulfophenyl diazenyl phenyl diazenyl-, sodium salt 1:6, hexasodium 4-hydroxy-7-5-hydroxy-7-sulfonato-6-2-2-sulfonato-4-2-4-sulfonatophenyl diazen-1-yl phenyl diazen-1-yl naphthalen-2-yl carbamoyl amino-3-2-2-sulfonato-4-2-4-sulfonatophenyl diazen-1-yl phenyl diazen-1-yl naphthalene-2-sulfonate
PubChem CID 57369623
IUPAC Name hexasodium;4-oxo-7-[[5-oxo-7-sulfonato-6-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-yl]carbamoylamino]-3-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2-sulfonate
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C(=CC4=C(C3=O)C=CC(=C4)NC(=O)NC5=CC6=C(C=C5)C(=O)C(=NNC7=C(C=C(C=C7)N=NC8=CC=C(C=C8)S(=O)(=O)[O-])S(=O)(=O)[O-])C(=C6)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 5 g
Formula Weight 1373.05
Physical Form Crystalline Powder
Chemical Name or Material Direct Red 80 [for Biochemical Research]
EINECSNumber (5)-1247
TSCA Yes