missing translation for 'onlineSavingsMsg'
Learn More

Direct Red 80, TCI America™

Catalog No. D327325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D327325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D327325G Supplier TCI America Supplier No. D327325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2610-10-8
Molecular Formula C45H26N10Na6O21S6
Molecular Weight (g/mol) 1373.05
MDL Number MFCD00054389
InChI Key LDCKXVAPQFUIOI-ZFLDVXHKSA-H
Synonym Sirius Red
PubChem CID 57369623
IUPAC Name hexasodium;4-oxo-7-[[5-oxo-7-sulfonato-6-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-yl]carbamoylamino]-3-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2-sulfonate
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C(=CC4=C(C3=O)C=CC(=C4)NC(=O)NC5=CC6=C(C=C5)C(=O)C(=NNC7=C(C=C(C=C7)N=NC8=CC=C(C=C8)S(=O)(=O)[O-])S(=O)(=O)[O-])C(=C6)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 1373.05
Chemical Name or Material Direct Red 80
EINECSNumber (5)-1247
TSCA Yes