accessibility menu, dialog, popup

UserName

NaNaH3C(S)O(R)POOO

CAS RN 26016-99-9

IUPAC Name: disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
Synonyms: fosfomycin sodium
Molecular Weight (g/mol): 182.02
Molecular Formula: C3H5Na2O4P
InChi Key: QZIQJIKUVJMTDG-JSTPYPERSA-L
SMILES: [Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
Color 
  • (4)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)