accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Cell Culture Media, Supplements, and Reagents
Cell Culture Reagents, Antibiotics and Supplements
Antibiotics
ICN15187601
Structure Search
Print
Phosphomycin Disodium Salt, MP Biomedicals
Catalog No.
ICN15187601
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
ICN15187601
1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
ICN15187601
Supplier
MP Biomedicals Inc
Supplier No.
0215187601
Sign In
or
Register
to check your price and availability.
$65.68
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
26016-99-9
Molecular Formula
C3H5Na2O4P
Molecular Weight (g/mol)
182.02
MDL Number
MFCD00056853
InChI Key
QZIQJIKUVJMTDG-JSTPYPERSA-L
PubChem CID
16397688
IUPAC Name
disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
SMILES
[Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O
Specifications
Quantity
1 g
Product Suggestions
Documents