accessibility menu, dialog, popup

UserName

CAS RN 259807-97-1

CH3SnH3CCH3NCH3

CAS RN 259807-97-1

IUPAC Name: 3-methyl-2-(tributylstannyl)pyridine
Synonyms: 3-methyl-2-(tributylstannyl)pyridine
Molecular Weight (g/mol): 382.18
Molecular Formula: C18H33NSn
InChi Key: QWDWZQDTTVYOBK-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=C(C)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results