missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-(tri-n-butylstannyl)pyridine, 96%
Description
3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.
Solubility
Sparingly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
3-Methyl-2-(tri-n-butylstannyl)pyridine is used as pharmaceutical intermediate.
Solubility
Sparingly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications
Specifications
| Quantity | 250 mg |
| UN Number | UN2788 |
| Solubility Information | Sparingly soluble in water. |
| Formula Weight | 382.16 |
| Percent Purity | 96% |
| Chemical Name or Material | 3-Methyl-2-(tri-n-butylstannyl)pyridine |
Safety and Handling
| Hazard Category | H301-H312-H315-H319-H372 |
| Hazard Statement | GHS H Statement H301-H372-H312-H315-H319 Toxic if swallowed. Causes damage to organs through prolonged or repeated exposure. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only