accessibility menu, dialog, popup

UserName

NN(?)HClNCH3MeMe

CAS RN 25952-53-8

IUPAC Name: ({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
Synonyms: ({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
Molecular Weight (g/mol): 191.7
Molecular Formula: C8H18ClN3
InChi Key: FPQQSJJWHUJYPU-UHFFFAOYSA-N
SMILES: Cl.CCN=C=NCCCN(C)C
Molecular Weight (g/mol) 
  • (11)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (8)
  • (1)
Grade 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (1)
  • (1)
  • (3)
  • (8)
  • (11)
  • (1)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (8)
  • (1)
  • (3)
  • (1)