missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide Hydrochloride 98.0+%, TCI America™

Catalog No. D16015G
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16015G 5 g
D160125G 25 g
D1601250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D16015G Supplier TCI America Supplier No. D16015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25952-53-8
Molecular Formula C8H18ClN3
Molecular Weight (g/mol) 191.70
MDL Number MFCD00012503
InChI Key FPQQSJJWHUJYPU-UHFFFAOYSA-N
Synonym 1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl
PubChem CID 2723939
IUPAC Name ({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
SMILES Cl.CCN=C=NCCCN(C)C

Specifications

Melting Point 115°C
Color White
pH 6.0 - 8.0 (50g/L)
Quantity 5 g
Formula Weight 191.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide Hydrochloride [Coupling Agent for Peptides Synthesis]
EINECSNumber (2)-1696
RTECSNumber FF2200000
TSCA Yes
Recommended Storage Refrigerator