missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%

Catalog No. AAA1080714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1080714 25 g
AAA1080703 1 g
AAA1080706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1080714 Supplier Thermo Scientific Chemicals Supplier No. A1080714

missing translation for 'validMainframeMsg'

CAS: 25952-53-8 | C8H18ClN3 | 191.70 g/mol

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride is used as a carboxyl activating agent. It is used in peptide synthesis, 3'-amino-3'-deoxyadenosine-5'-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride is used as a carboxyl activating agent. It is used in peptide synthesis, 3′-amino-3′-deoxyadenosine-5′-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

Solubility
Soluble in acetonitrile dichloromethane, dimethylformamide, and tetrahydrofuran. Slightly soluble in water.

Notes
Sensitive to moisture. Incompatible with strong acids, strong oxidizing agents and moisture.

Chemical Identifiers

CAS 25952-53-8
Molecular Formula C8H18ClN3
Molecular Weight (g/mol) 191.70
MDL Number MFCD00012503
InChI Key FPQQSJJWHUJYPU-UHFFFAOYSA-N
Synonym 1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride, edc.hcl, edci, edc hydrochloride, wsc hcl, edac hydrochloride, edac, hydrochloride, 1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride, edcl, edac hcl
PubChem CID 2723939
IUPAC Name 3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CCN=C=NCCCN(C)C

Specifications

Melting Point ∼113°C
Linear Formula CH3CH2N=C=N(CH2)3N(CH3)2·HCl
Quantity 25 g
Beilstein 5764110
Sensitivity Moisture sensitive
Solubility Information Soluble in acetonitrile dichloromethane,dimethylformamide,and tetrahydrofuran. Slightly soluble in water.
Formula Weight 191.7
Percent Purity ≥98%
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide Hydrochloride
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 247-361-2
RTECSNumber FF2200000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only