accessibility menu, dialog, popup

UserName

CH3CH3H3COOHN(S)H3NOO

CAS RN 25691-37-6

IUPAC Name: (2S)-4-azaniumyl-2-{[(tert-butoxy)carbonyl]amino}butanoate
Synonyms: (2S)-4-ammonio-2-[(tert-butoxycarbonyl)amino]butanoate
Molecular Weight (g/mol): 218.25
Molecular Formula: C9H18N2O4
InChi Key: MDCPCLPRWLKUIQ-LURJTMIESA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC[NH3+])C([O-])=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)