missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Amino-2-(Boc-amino)butyric acid, 97%

Catalog No. AAH6245006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6245006 5 g
AAH6245003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6245006 Supplier Thermo Scientific Chemicals Supplier No. H6245006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25691-37-6 | C9H18N2O4 | 218.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25691-37-6
Molecular Formula C9H18N2O4
Molecular Weight (g/mol) 218.253
MDL Number MFCD00236841
InChI Key MDCPCLPRWLKUIQ-LURJTMIESA-N
Synonym boc-l-2,4-diaminobutyric acid, boc-dab-oh, s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, s-4-amino-2-tert-butoxycarbonylamino butanoic acid, n2-boc-s-2,4-diamino-butanoic acid, s-4-amino-2-boc-amino butyric acid, 2s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, n-boc-s-2,4-diamino-butanoic acid, butanoic acid, 4-amino-2-1,1-dimethylethoxy carbonyl amino-, 2s, s-4-amino-2-tert-butoxycarbonylamino butyric acid
PubChem CID 7019666
IUPAC Name (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NC(CCN)C(=O)O

Specifications

Color White
Quantity 5 g
Beilstein 4745487
Formula Weight 218.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-4-Amino-2-(Boc-amino)butyric acid
Hazard Category H315-H318
Hazard Statement GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only