accessibility menu, dialog, popup

UserName

H2COON

CAS RN 25635-24-9

IUPAC Name: prop-2-en-1-yl pyridine-4-carboxylate
Synonyms: prop-2-en-1-yl pyridine-4-carboxylate
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: ORSMQFTVFDSEBA-UHFFFAOYSA-N
SMILES: C=CCOC(=O)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results