missing translation for 'onlineSavingsMsg'
Learn More

Allyl Isonicotinate 98.0+%, TCI America™

Catalog No. I013625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I013625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I013625ML Supplier TCI America Supplier No. I013625ML

Chemical Identifiers

CAS 25635-24-9
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00059786
InChI Key ORSMQFTVFDSEBA-UHFFFAOYSA-N
Synonym Isonicotinic Acid Allyl Ester, Allyl 4-Pyridinecarboxylate, 4-Pyridinecarboxylic Acid Allyl Ester
PubChem CID 571571
IUPAC Name prop-2-enyl pyridine-4-carboxylate
SMILES C=CCOC(=O)C1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 106°C
Quantity 25 mL
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Isonicotinate