missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperidine-4-carboxylic acid, 98+%

Catalog No. AAA1560622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1560622 100 g
AAA1560606 5 g
AAA1560614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1560622 Supplier Thermo Scientific Chemicals Supplier No. A1560622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25503-90-6 | C8H12NO3 | 170.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25503-90-6
Molecular Formula C8H12NO3
Molecular Weight (g/mol) 170.19
MDL Number MFCD00023706
InChI Key WFCLWJHOKCQYOQ-UHFFFAOYSA-M
Synonym 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl
PubChem CID 117255
IUPAC Name 1-acetylpiperidine-4-carboxylic acid
SMILES CC(=O)N1CCC(CC1)C([O-])=O

Specifications

Melting Point 180°C to 184°C
Quantity 100 g
Beilstein 389577
Formula Weight 171.19
Percent Purity ≥98%
Chemical Name or Material 1-Acetylpiperidine-4-carboxylic acid
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 247-049-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only