missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. A22615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22615G 5 g
A226125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22615G Supplier TCI America Supplier No. A22615G

Chemical Identifiers

CAS 25503-90-6
Molecular Formula C8H12NO3
Molecular Weight (g/mol) 170.19
MDL Number MFCD00023706
InChI Key WFCLWJHOKCQYOQ-UHFFFAOYSA-M
Synonym 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl
PubChem CID 117255
IUPAC Name 1-acetylpiperidine-4-carboxylate
SMILES CC(=O)N1CCC(CC1)C([O-])=O

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 5 g
Formula Weight 171.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetyl-4-piperidinecarboxylic Acid