accessibility menu, dialog, popup

UserName

CAS RN 2548-87-0

H3CO(E)

CAS RN 2548-87-0

IUPAC Name: (2E)-oct-2-enal
Synonyms: (E)-2-octenal trans-octen-2-al octene-1-oxide
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: LVBXEMGDVWVTGY-VOTSOKGWSA-N
SMILES: CCCCC\C=C\C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results