missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octenal 96.0+%, TCI America™

Catalog No. O017625ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O017625ML 25 mL
O017610ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O017625ML Supplier TCI America Supplier No. O017625ML

Chemical Identifiers

CAS 2548-87-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00007011
InChI Key LVBXEMGDVWVTGY-VOTSOKGWSA-N
Synonym trans-2-octenal, e-oct-2-enal, e-2-octenal, 2-octenal, trans-2-octen-1-al, e-2-octen-1-al, 2-octen-1-al, 2-octenel, 2-octenal, e, 2-octenal, 2e
PubChem CID 5283324
ChEBI CHEBI:61748
IUPAC Name (E)-oct-2-enal
SMILES CCCCCC=CC=O

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 mL
Formula Weight 126.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Octenal
RTECSNumber RH2145000
TSCA Yes