CAS RN 25393-98-0
IUPAC Name:
1,4-bis(1,2-dibromoethyl)benzene
Synonyms:
1,4-bis(1,2-dibromoethyl)benzene
Molecular Weight (g/mol):
449.81
Molecular Formula:
C10H10Br4
InChi Key:
KDBQQANTDCVPDZ-UHFFFAOYNA-N
SMILES:
BrCC(Br)C1=CC=C(C=C1)C(Br)CBr
1
–
1
of
1
results
1
1,4-Bis(1,2-dibromoethyl)benzene 97.0+%, TCI America™
CAS: 25393-98-0 Molecular Formula: C10H10Br4 Molecular Weight (g/mol): 449.81 MDL Number: MFCD00017881 InChI Key: KDBQQANTDCVPDZ-UHFFFAOYNA-N Synonym: alpha,alpha′C,beta,beta′C-Tetrabromo-1,4-diethylbenzene PubChem CID: 97018 IUPAC Name: 1,4-bis(1,2-dibromoethyl)benzene SMILES: BrCC(Br)C1=CC=C(C=C1)C(Br)CBr