missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(1,2-dibromoethyl)benzene 97.0+%, TCI America™

Catalog No. B15045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15045G Supplier TCI America Supplier No. B15045G

Chemical Identifiers

CAS 25393-98-0
Molecular Formula C10H10Br4
Molecular Weight (g/mol) 449.81
MDL Number MFCD00017881
InChI Key KDBQQANTDCVPDZ-UHFFFAOYNA-N
Synonym alpha,alpha′C,beta,beta′C-Tetrabromo-1,4-diethylbenzene
PubChem CID 97018
IUPAC Name 1,4-bis(1,2-dibromoethyl)benzene
SMILES BrCC(Br)C1=CC=C(C=C1)C(Br)CBr

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 449.81
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(1,2-dibromoethyl)benzene
RTECSNumber CY8440000
TSCA No