accessibility menu, dialog, popup

UserName

H3CCH3HOH3CCH3OOCH3H3C

CAS RN 25265-77-4

IUPAC Name: 3-hydroxy-2,2,4-trimethylpentyl 2-methylpropanoate
Synonyms: texanol trimethyl hydroxypentyl isobutyrate
Molecular Weight (g/mol): 216.32
Molecular Formula: C12H24O3
InChi Key: DAFHKNAQFPVRKR-UHFFFAOYNA-N
SMILES: CC(C)C(O)C(C)(C)COC(=O)C(C)C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)