accessibility menu, dialog, popup

UserName

CAS RN 25213-24-5

CH3OOH2CH2COH

CAS RN 25213-24-5

IUPAC Name: ethenol; ethenyl acetate
Synonyms: vinyl acetate; vinyl alcohol
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: CALWOYBZYFNRDN-UHFFFAOYSA-N
SMILES: OC=C.CC(=O)OC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results