missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinyl Alcohol, Partially Hydrolyzed, MilliporeSigma™
Specifications
Specifications
| Melting Point | >200°C |
| Color | Colorless |
| Density | 1.3g/cm3 |
| Ignition Point | >230°C (>446°F) |
| Odor | Odorless |
| Packaging | Plastic Bottle |
| Quantity | 1 kg |
| Solubility Information | Soluble in hot water, at 20°C insoluble |
| Physical Form | Solid |
| Chemical Name or Material | Polyvinyl Alcohol |
Safety and Handling
| Hazard Category | H370: Causes damage to organs (Eyes). |
| Recommended Storage | 15° to 25°C (59° to 77°F); Tightly closed. Dry. |