accessibility menu, dialog, popup

UserName

∗∗Nn

CAS RN 25067-59-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C14H11N)n
InChi Key: —
SMILES: *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results