missing translation for 'onlineSavingsMsg'
Learn More

Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400

Catalog No. AC329690250
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC329690250 25 g
AC329690050 5 g
AC179030100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC329690250 Supplier Thermo Scientific Chemicals Supplier No. 329690250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25067-59-8 | (C14H11N)n | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25067-59-8
Molecular Formula (C14H11N)n
Molecular Weight (g/mol) 193.25
MDL Number MFCD00134336
InChI Key KKFHAJHLJHVUDM-UHFFFAOYSA-N
Synonym 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole
PubChem CID 15143
SMILES *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White
Physical Form Crystalline Powder
Density 1.2000g/mL
Quantity 25 g
Infrared Spectrum Authentic
Loss on Drying 3% max.
Packaging Glass bottle
Refractive Index 1.683
Specific Gravity 1.2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, dichloromethane,, dioxane, thf, toluene
IUPAC Name 9-ethenylcarbazole
Chemical Name or Material Poly(N-vinylcarbazole), Secondary standard
Show More Show Less
RTECSNumber FE6225480
TSCA TSCA

RUO – Research Use Only