accessibility menu, dialog, popup

UserName

CH3H3CCH3NOH3CH3CCH3CH3H3CCH3

CAS RN 24973-59-9

IUPAC Name: 1,3,5-tri-tert-butyl-2-nitrosobenzene
Synonyms: 1,3,5-tri-tert-butyl-2-nitrosobenzene
Molecular Weight (g/mol): 275.44
Molecular Formula: C18H29NO
InChi Key: OSICDPWAPKXXHT-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=C(N=O)C(=C1)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results