accessibility menu, dialog, popup

UserName

CAS RN 24932-48-7

IUPAC Name: 1,2-dibromo-4,5-dimethylbenzene
Synonyms: 1,2-dibromo-4,5-dimethylbenzene
Molecular Weight (g/mol): 263.96
Molecular Formula: C8H8Br2
InChi Key: BCIDDURGCAHERU-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(Br)C=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)