accessibility menu, dialog, popup

UserName

CAS RN 24851-98-7

IUPAC Name: methyl 2-(3-oxo-2-pentylcyclopentyl)acetate
Synonyms: methyl dihydrojasmonate sodium tetrahydrojasmonate
Molecular Weight (g/mol): 226.32
Molecular Formula: C13H22O3
InChi Key: KVWWIYGFBYDJQC-UHFFFAOYNA-N
SMILES: CCCCCC1C(CC(=O)OC)CCC1=O
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results