missing translation for 'onlineSavingsMsg'
Learn More

Methyl dihydrojasmonate, mixture of cis and trans, ≥96% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013693
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013693 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013693 Supplier Merck Emd Millipore Supplier No. 596771ML

Specifications

CAS 24851-98-7
Density 0.998 g/mL (at 25°C)
Flash Point 113°C - closed cup
Linear Formula C13H22O3
MDL Number MFCD00085362
Boiling Point 110°C (0.2 mmHg)
Synonym (3-Oxo-2-pentylcyclopentyl)acetic acid methyl ester; Methyl (3-oxo-2-pentylcyclopentyl)acetate
Type Analytical Standard
Quantity 1 mL
Formula Weight 226.31
Percent Purity ≥96% (GC)
Grade Analytical Standard
Show More Show Less