accessibility menu, dialog, popup

UserName

H3CONH2OOCH3

CAS RN 2475-80-1

IUPAC Name: methyl 2-amino-5-methoxybenzoate
Synonyms: methyl 2-amino-5-methoxybenzoate
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: MOVBJUGHBJJKOW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=CC(OC)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results