accessibility menu, dialog, popup

UserName

H3CNHHNCH3OO

CAS RN 2475-44-7

IUPAC Name: 1,4-bis(methylamino)-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,4-bis(methylamino)anthracene-9,10-dione
Molecular Weight (g/mol): 266.3
Molecular Formula: C16H14N2O2
InChi Key: QOSTVEDABRQTSU-UHFFFAOYSA-N
SMILES: CNC1=CC=C(NC)C2=C1C(=O)C1=CC=CC=C1C2=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results