missing translation for 'onlineSavingsMsg'
Learn More

Disperse Blue 14, TCI America™

Catalog No. D328025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D328025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D328025G Supplier TCI America Supplier No. D328025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2475-44-7
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.30
MDL Number MFCD00001198
InChI Key QOSTVEDABRQTSU-UHFFFAOYSA-N
Synonym disperse blue 14, 1,4-bis methylamino anthraquinone, resiren blue tb, solvent blue 78, acetate blue b, celliton fast blue b, cibacet blue br, setacyl blue bs, teraprint blue r, 1,4-bis methylamino anthracene-9,10-dione
PubChem CID 17189
IUPAC Name 1,4-bis(methylamino)-9,10-dihydroanthracene-9,10-dione
SMILES CNC1=CC=C(NC)C2=C1C(=O)C1=CC=CC=C1C2=O

Specifications

Melting Point 222°C
Color Purple
Quantity 25 g
Formula Weight 266.30
Physical Form Crystalline Powder
Chemical Name or Material Disperse Blue 14
EINECSNumber (5)-2572&(5)-5119
RTECSNumber CB5750000
TSCA Yes