accessibility menu, dialog, popup

UserName

HO

CAS RN 24595-58-2

IUPAC Name: 5-phenylpent-4-yn-1-ol
Synonyms: 5-phenylpent-4-yn-1-ol
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: QQBCRIBCSOQZFZ-UHFFFAOYSA-N
SMILES: OCCCC#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results