missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-4-pentyn-1-ol 98.0+%, TCI America™

Catalog No. P11581G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11581G Supplier TCI America Supplier No. P11581G
Only null left

Chemical Identifiers

CAS 24595-58-2
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.22
MDL Number MFCD00191481
InChI Key QQBCRIBCSOQZFZ-UHFFFAOYSA-N
PubChem CID 11137434
IUPAC Name 5-phenylpent-4-yn-1-ol
SMILES OCCCC#CC1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 160.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Phenyl-4-pentyn-1-ol