accessibility menu, dialog, popup

UserName

FFFNHON

CAS RN 24522-30-3

IUPAC Name: 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Synonyms: 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Molecular Weight (g/mol): 228.17
Molecular Formula: C10H7F3N2O
InChi Key: JBNCFFDGYDZEEN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(NC(=O)CC#N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results