CAS RN 24502-79-2
CAS RN 24502-79-2
IUPAC Name:
1-(5,8-dihydroxy-1,4-dioxo-1,4-dihydronaphthalen-2-yl)-4-methylpent-3-en-1-yl 3-methylbut-2-enoate
Synonyms:
1-(5,8-dihydroxy-1,4-dioxonaphthalen-2-yl)-4-methylpent-3-en-1-yl 3-methylbut-2-enoate
Molecular Weight (g/mol):
370.4
Molecular Formula:
C21H22O6
InChi Key:
BATBOVZTQBLKIL-UHFFFAOYNA-N
SMILES:
CC(C)=CCC(OC(=O)C=C(C)C)C1=CC(=O)C2=C(O)C=CC(O)=C2C1=O
1
–
1
of
1
results
1
(beta,beta-Dimethylacryl)shikonin 90.0+%, TCI America™
CAS: 24502-79-2 Molecular Formula: C21H22O6 Molecular Weight (g/mol): 370.401 MDL Number: MFCD00143687 InChI Key: BATBOVZTQBLKIL-QGZVFWFLSA-N Synonym: 5,8-Dihydroxy-2-[1-(beta,beta-dimethylacryloyloxy)-4-methyl-3-pentenyl]-1,4-naphthalenedione PubChem CID: 479499 ChEBI: CHEBI:81071 IUPAC Name: [(1R)-1-(5,8-dihydroxy-1,4-dioxonaphthalen-2-yl)-4-methylpent-3-enyl] 3-methylbut-2-enoate SMILES: CC(=CCC(C1=CC(=O)C2=C(C=CC(=C2C1=O)O)O)OC(=O)C=C(C)C)C