accessibility menu, dialog, popup

UserName

HOONPHOOHOPOHHOO

CAS RN 2439-99-8

IUPAC Name: 2-[bis(phosphonomethyl)amino]acetic acid
Synonyms: glyphosine glyphosphine
Molecular Weight (g/mol): 263.08
Molecular Formula: C4H11NO8P2
InChi Key: OXHDYFKENBXUEM-UHFFFAOYSA-N
SMILES: OC(=O)CN(CP(O)(O)=O)CP(O)(O)=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results