missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Glyphosine, ≥98%, Calbiochem™,

Catalog No. 3567905GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3567905GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3567905GM Supplier MilliporeSigma™ Supplier No. 3567905GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2439-99-8
Molecular Formula C4H11NO8P2
Molecular Weight (g/mol) 263.079
InChI Key OXHDYFKENBXUEM-UHFFFAOYSA-N
Synonym glyphosine, n,n-bis phosphonomethyl glycine, glyphosphine, polaris, 2-bis phosphonomethyl amino acetic acid, glycine, n,n-bis phosphonomethyl, glyphosine ansi, caswell no. 471a, unii-2u96121skr
PubChem CID 17112
ChEBI CHEBI:63484
IUPAC Name 2-[bis(phosphonomethyl)amino]acetic acid
SMILES C(C(=O)O)N(CP(=O)(O)O)CP(=O)(O)O

Specifications

Color White
Packaging Glass Bottle
Quantity 5 g
Percent Purity ≥98% (NMR)
Physical Form Solid