missing translation for 'onlineSavingsMsg'
Learn More

Glycine-N,N-bis(methylenephosphonic Acid) 97.0+%, TCI America™

Catalog No. G02295G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G02295G Supplier TCI America Supplier No. G02295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2439-99-8
Molecular Formula C4H11NO8P2
Molecular Weight (g/mol) 263.079
MDL Number MFCD00056670
InChI Key OXHDYFKENBXUEM-UHFFFAOYSA-N
Synonym glyphosine, n,n-bis phosphonomethyl glycine, glyphosphine, polaris, 2-bis phosphonomethyl amino acetic acid, glycine, n,n-bis phosphonomethyl, glyphosine ansi, caswell no. 471a, unii-2u96121skr
PubChem CID 17112
ChEBI CHEBI:63484
IUPAC Name 2-[bis(phosphonomethyl)amino]acetic acid
SMILES C(C(=O)O)N(CP(=O)(O)O)CP(=O)(O)O

Specifications

Color White
Quantity 5 g
Formula Weight 263.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycine-N,N-bis(methylenephosphonic Acid)
RTECSNumber MB9120000
TSCA Yes