missing translation for 'onlineSavingsMsg'
Learn More

(+)-Terpinen-4-ol, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111018891
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018891 1 mL
111018233 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018891 Supplier Merck Emd Millipore Supplier No. 864771ML

(+)-Terpinen-4-ol is an important constituent of tea tree oil, possessing antimycotic activity, which can be extracted from the leaves and flowers of Melaleuca alternifolia.

Featured Industries: Food and Beverages

Specifications

CAS 2438-10-0
Density 0.933 g/mL (at 20°C)
Flash Point 79°C - closed cup
Physical Form Neat
Linear Formula C10H18 O
MDL Number MFCD00077431
Boiling Point 211°C to 213°C
Synonym (S)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol; (S)-p-Menth-1-en-4-ol
Type Analytical Standard
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less