accessibility menu, dialog, popup

UserName

CAS RN 2438-04-2

IUPAC Name: 2-(propan-2-yl)benzoic acid
Synonyms: O-isopropylbenzoic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: BANZVKGLDQDFDV-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results