accessibility menu, dialog, popup

UserName

CAS RN 2437-56-1

IUPAC Name: tridec-1-ene
Synonyms: 1-tridecene
Molecular Weight (g/mol): 182.35
Molecular Formula: C13H26
InChi Key: VQOXUMQBYILCKR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)