missing translation for 'onlineSavingsMsg'
Learn More

1-Tridecene 99.5+%, TCI America™

Catalog No. S03435ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03435ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03435ML Supplier TCI America Supplier No. S03435ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2437-56-1
Molecular Formula C13H26
Molecular Weight (g/mol) 182.351
MDL Number MFCD00008976
InChI Key VQOXUMQBYILCKR-UHFFFAOYSA-N
Synonym 1-tridecene, n-tridec-1-ene, tridecylene, alpha-tridecene, undecylethylene, tridecene-1, unii-5b0u2s314c, ccris 5719, 1-tridecylene, .alpha.-tridecene
PubChem CID 17095
IUPAC Name tridec-1-ene
SMILES CCCCCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 233°C
Quantity 5 mL
Formula Weight 182.35
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material 1-Tridecene [Standard Material for GC]
EINECSNumber (2)-0027
RTECSNumber YD4595000
TSCA Yes